Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-Pyrrolidone-5-carboxylic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Plastic bottle
98-79-3
C5H7NO3
129.12
ODHCTXKNWHHXJC-VKHMYHEASA-N
l-pyroglutamic acid,pyroglutamic acid,pidolic acid,5-oxo-l-proline,h-pyr-oh,5-oxoproline,s-5-oxopyrrolidine-2-carboxylic acid,2s-5-oxopyrrolidine-2-carboxylic acid,l-5-oxoproline,l-proline, 5-oxo
7405
CHEBI:18183
(2S)-5-oxopyrrolidine-2-carboxylic acid
C1CC(=O)NC1C(=O)O
98-79-3
C5H7NO3
129.12
ODHCTXKNWHHXJC-VKHMYHEASA-N
l-pyroglutamic acid,pyroglutamic acid,pidolic acid,5-oxo-l-proline,h-pyr-oh,5-oxoproline,s-5-oxopyrrolidine-2-carboxylic acid,2s-5-oxopyrrolidine-2-carboxylic acid,l-5-oxoproline,l-proline, 5-oxo
7405
CHEBI:18183
(2S)-5-oxopyrrolidine-2-carboxylic acid
C1CC(=O)NC1C(=O)O
Specific Rotation Condition − 27.50 (20.00°C c=10,1 N NaOH)
Water 0.5% Max. (K.F.)
Specific Rotation − 27.50
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 100g
Melting Point 152.0°C to 162.0°C
Color White
Assay Percent Range 98%
Beilstein 22,284
Merck Index 15,8113
Solubility Information Solubility in water: 100-150g/l (20°). Other solubilities: soluble in alcohol and acetone
Formula Weight 129.12
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material (S)-(-)-2-Pyrrolidone-5-carboxylic acid
Show More Show Less

GHS P Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.