Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
66605-57-0
C14H21NO3
251.326
MFCD00076976
LDKDMDVMMCXTMO-LBPRGKRZSA-N
n-boc-l-phenylalaninol,boc-l-phenylalaninol,boc-phenylalaninol,s-2-boc-amino-3-phenyl-1-propanol,n-tert-butoxycarbonyl-l-phenylalaninol,tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate,s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate,n-boc-l-phenylalaninol,s-n-tert-butoxycarbonyl-phenylalaninol,s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
2733675
tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CO
66605-57-0
C14H21NO3
251.326
MFCD00076976
LDKDMDVMMCXTMO-LBPRGKRZSA-N
n-boc-l-phenylalaninol,boc-l-phenylalaninol,boc-phenylalaninol,s-2-boc-amino-3-phenyl-1-propanol,n-tert-butoxycarbonyl-l-phenylalaninol,tert-butyl n-2s-1-hydroxy-3-phenylpropan-2-yl carbamate,s-tert-butyl 1-hydroxy-3-phenylpropan-2-yl carbamate,n-boc-l-phenylalaninol,s-n-tert-butoxycarbonyl-phenylalaninol,s---2-tert-butoxycarbonylamino-3-phenyl-1-propanol
2733675
tert-butyl N-[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamate
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)CO
Melting Point >94°C
Color White
Specific Rotation -22° to -28° (20°C, 589nm) (c=1, CHCl3)
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 251.32
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material (S)-(-)-2-(tert-Butoxycarbonylamino)-3-phenyl-1-propanol
Show More Show Less