Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Trifluoromethyl)piperidine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
154727-51-2
C6H10F3N
153.15
NAXDEFXCCITWEU-YFKPBYRVSA-N
s-2-trifluoromethyl piperidine,2s-2-trifluoromethyl piperidine,s-2-trifluoromethylpiperidine,2alpha-trifluoromethyl piperidine
42580051
(2S)-2-(trifluoromethyl)piperidine
C1CCNC(C1)C(F)(F)F
154727-51-2
C6H10F3N
153.15
NAXDEFXCCITWEU-YFKPBYRVSA-N
s-2-trifluoromethyl piperidine,2s-2-trifluoromethyl piperidine,s-2-trifluoromethylpiperidine,2alpha-trifluoromethyl piperidine
42580051
(2S)-2-(trifluoromethyl)piperidine
C1CCNC(C1)C(F)(F)F
Quantity 5g
Boiling Point 80.0°C (200.0 Torr)
Assay Percent Range 96% min. (GC)
Formula Weight 153.15
Percent Purity 97%
Chemical Name or Material (S)-2-(Trifluoromethyl)piperidine

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger