Learn More
Tocris Bioscience™ S 2101 Available on GSA/VA Contract for Federal Government customers only.
LSD1 inhibitor
$196.00 - $823.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
571410 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5714/10
10mg Each for $196.00
Add to cart
571450 View Documents Tocris Bioscience™
5714/50
Each for $823.00
Add to cart
Description
Description
LSD1 inhibitor (IC50 = 990 nM; Ki = 610 nM). Exhibits selectivity for LSD1 over MAO-B and MAO-A (Ki values are 17 and 110 μM, respectively).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Safety and Handling
Safety and Handling
Recommended Storage : Store at 4°C