Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-3,5-DHPG, Tocris Bioscience™
Click to view available options
Quantity:
5 mg
10 mg
Description
(S)-3,5-DHPG is a selective group I mGlu receptor agonist. (S)-3,5-DHPG regulates the expression of microRNAs in mouse cerebral cortex in vivo. (S)-3,5-DHPG induces a profound depression of synaptic dopamine release in mouse cortico-striatal cells in vitro, and anxiety-like behavior in vivo in mice.
Racemate also available.
Specifications
Formula Weight | 183.16 |
Percent Purity | >98% |
Quantity | 5 mg |
Chemical Name or Material | (S)-3, 5-DHPG |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction