Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3,5-DHPG, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
5mg
10mg

Chemical Identifiers

162870-29-3
C8H9NO4
183.163
HOOWCUZPEFNHDT-ZETCQYMHSA-N
(S)-3, 5-Dihydroxyphenylglycine
443586
CHEBI:29474

Selective group I mGlu agonist. Active enantiomer of 3,5-DHPG (Cat. No. 0342)

Selective group I mGlu receptor agonist. Also available as part of the Group I mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . (RS)-3,5-DHPG (Cat. No. 0342) also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

SMILES C1=C(C=C(C=C1O)O)C(C(=O)O)N
IUPAC Name (2S)-2-amino-2-(3,5-dihydroxyphenyl)acetic acid
Formula Weight 183.16
Quantity 10mg
Chemical Name or Material (S)-3, 5-DHPG
Grade >98%

Recommended Storage : Desiccate at -20°C