Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-(Acetylthio)-2-methylpropionic Acid 98.0+%, TCI America™

Click to view available options
Quantity:
5mL
25mL
76497-39-7
C6H10O3S
162.203
MFCD00038563
VFVHNRJEYQGRGE-SCSAIBSYSA-N
2724506
(2S)-3-acetylsulfanyl-2-methylpropanoic acid
CC(CSC(=O)C)C(=O)O
76497-39-7
C6H10O3S
162.203
MFCD00038563
VFVHNRJEYQGRGE-SCSAIBSYSA-N
2724506
(2S)-3-acetylsulfanyl-2-methylpropanoic acid
CC(CSC(=O)C)C(=O)O
Quantity 5mL
Color Yellow
Melting Point 5°C
Boiling Point 132°C
Formula Weight 162.20
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (T)
Chemical Name or Material (S)-(-)-3-(Acetylthio)-2-methylpropionic Acid

EINECSNumber : (2)-3239

TSCA : No