Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Amino-1,2-propanediol, 98%, Thermo Scientific Chemicals

Catalog Number aah2674303
Click to view available options
Quantity:
1 g
5 g
61278-21-5
C3H9NO2
91.11
MFCD00798260
KQIGMPWTAHJUMN-VKHMYHEASA-N
s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
440902
CHEBI:1456
(2S)-3-aminopropane-1,2-diol
C(C(CO)O)N
61278-21-5
C3H9NO2
91.11
MFCD00798260
KQIGMPWTAHJUMN-VKHMYHEASA-N
s-3-amino-1,2-propanediol, s-3-aminopropane-1,2-diol, s---3-amino-1,2-propanediol, 2s-3-aminopropane-1,2-diol, s---3-amino-1,2-propandiol, 1,2-propanediol, 3-amino-, 2s, ksc496i7p, s-2,3-dihydroxypropylamine, 2s-2,3-dihydroxypropylamine, 2s-3-amino-1,2-propanediol
440902
CHEBI:1456
(2S)-3-aminopropane-1,2-diol
C(C(CO)O)N

(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-3-Amino-1,2-propanediol is used as a chiral selector ligand, used subsequently for a nucleophilic substitution with the dimethoxytrityl (DMT)-protected

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 54°C to 56°C
Density 1.175
Boiling Point 117°C to 119°C (0.4 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.483
Quantity 1 g
UN Number UN3259
Beilstein 1719122
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation −28° (c=4, 5M HCl)
Formula Weight 91.11
Percent Purity 98%
Chemical Name or Material (S)-(-)-3-Amino-1,2-propanediol
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit