Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-3-Amino-1-BOC-piperidine, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
625471-18-3
C10H20N2O2
200.28
AKQXKEBCONUWCL-QMMMGPOBSA-N
s-1-boc-3-aminopiperidine,s-3-amino-1-n-boc-piperidine,tert-butyl 3s-3-aminopiperidine-1-carboxylate,s-tert-butyl 3-aminopiperidine-1-carboxylate,s-3-amino-1-boc-piperidine,s-+-3-amino-1-boc-piperidine,3s-3-aminopiperidine, n1-boc protected,s-1-boc-3-amniopiperidine,s-3-amino-piperidine-1-carboxylic acid tert-butyl ester,s-3-amino-n-tboc-piperidine
1501975
tert-butyl (3S)-3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N
625471-18-3
C10H20N2O2
200.28
AKQXKEBCONUWCL-QMMMGPOBSA-N
s-1-boc-3-aminopiperidine,s-3-amino-1-n-boc-piperidine,tert-butyl 3s-3-aminopiperidine-1-carboxylate,s-tert-butyl 3-aminopiperidine-1-carboxylate,s-3-amino-1-boc-piperidine,s-+-3-amino-1-boc-piperidine,3s-3-aminopiperidine, n1-boc protected,s-1-boc-3-amniopiperidine,s-3-amino-piperidine-1-carboxylic acid tert-butyl ester,s-3-amino-n-tboc-piperidine
1501975
tert-butyl (3S)-3-aminopiperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(C1)N
Specific Rotation 30
Quantity 5g
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Formula Weight 200.28
Physical Form Gum or Semi-Solid
Percent Purity 95%
Chemical Name or Material (S)-(+)-3-Amino-1-BOC-piperidine
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger