Alfa Aesar™ (S)-(+)-3-Amino-1-Boc-piperidine, 97%

Manufacturer: Alfa Aesar™ H30538MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH30538MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 625471-18-3
    Molecular Formula C10H20N2O2
    Formula Weight 200.28
    MDL Number MFCD03094718
    UN Number UN2735
    Synonym (S)-Boc-3-piperidinamine; tert-Butyl (R)-3-aminopiperidine-1-carboxylate
    Chemical Name or Material (S)-(+)-3-Amino-1-Boc-piperidine

    Specifications

    Formula Weight 200.28
    Molecular Formula C10H20N2O2
    UN Number UN2735
    Quantity 250mg
    MDL Number MFCD03094718
    CAS 625471-18-3
    Synonym (S)-Boc-3-piperidinamine; tert-Butyl (R)-3-aminopiperidine-1-carboxylate
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Chemical Name or Material (S)-(+)-3-Amino-1-Boc-piperidine
    Assay 97%

    Description

    Applications
    (S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

    Solubility
    Soluble in dimethylsulfoxide.

    Notes
    Air and carbon dioxide sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit