Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(+)-3-Aminotetrahydrofuran hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
204512-95-8
C4H10ClNO
123.58
MFCD08445642
MHOVLDXJDIEEMJ-WCCKRBBISA-N
s-tetrahydrofuran-3-amine hydrochloride,s-3-aminotetrahydrofuran hydrochloride,3s-oxolan-3-amine hydrochloride,s-3-aminotetrahydrofuran hcl,s-tetrahydrofuran-3-amine hcl,s-tetrahydro-furan-3-ylamine hydrochloride,3-furanamine, tetrahydro-, hydrochloride, 3s,s-3-aminotetrahydrofuranhydrochloride,pubchem15126,ksc916a5t
18664284
(3S)-oxolan-3-amine;hydrochloride
C1COCC1N.Cl
204512-95-8
C4H10ClNO
123.58
MFCD08445642
MHOVLDXJDIEEMJ-WCCKRBBISA-N
s-tetrahydrofuran-3-amine hydrochloride,s-3-aminotetrahydrofuran hydrochloride,3s-oxolan-3-amine hydrochloride,s-3-aminotetrahydrofuran hcl,s-tetrahydrofuran-3-amine hcl,s-tetrahydro-furan-3-ylamine hydrochloride,3-furanamine, tetrahydro-, hydrochloride, 3s,s-3-aminotetrahydrofuranhydrochloride,pubchem15126,ksc916a5t
18664284
(3S)-oxolan-3-amine;hydrochloride
C1COCC1N.Cl
Quantity 250mg
Beilstein 14637084
Formula Weight 123.58
Physical Form Solid
Percent Purity 95%
Chemical Name or Material (S)-(+)-3-Aminotetrahydrofuran hydrochloride

storageNote1 : -30° to -10°C