Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
104530-79-2
C7H8O3S·C4H9NO
259.32
MIPHRQMEIYLZFZ-BYPYZUCNSA-N
s-3-aminotetrahydrofuran,s-tetrahydrofuran-3-amine,3s-oxolan-3-amine,3-furanamine,tetrahydro-, 3s,s-tetrahydro-furan-3-ylamine,tetrahydrofuran-3beta-amine,s-3-amino-tetrahydrofuran,5-tetrahydrofuran-3-amine,3s-tetrahydro-3-furanamine,3s-tetrahydrofuran-3-amine
14243168
(3S)-oxolan-3-amine
C1COCC1N
104530-79-2
C7H8O3S·C4H9NO
259.32
MIPHRQMEIYLZFZ-BYPYZUCNSA-N
s-3-aminotetrahydrofuran,s-tetrahydrofuran-3-amine,3s-oxolan-3-amine,3-furanamine,tetrahydro-, 3s,s-tetrahydro-furan-3-ylamine,tetrahydrofuran-3beta-amine,s-3-amino-tetrahydrofuran,5-tetrahydrofuran-3-amine,3s-tetrahydro-3-furanamine,3s-tetrahydrofuran-3-amine
14243168
(3S)-oxolan-3-amine
C1COCC1N
Melting Point 137.9°C to 142.8°C
Specific Rotation -4
Infrared Spectrum Authentic
Color White to Brown
Quantity 1g
Packaging Glass bottle
Elemental Analysis 48.9 to 53.0 % C 5.2 to 5.6 % N
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 259.32
Physical Form Crystalline Powder or Flakes
Percent Purity 97%
Chemical Name or Material (S)-(-)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

GHS Signal Word: Warning