Alfa Aesar™ (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde, 95%

Manufacturer: Alfa Aesar™ H5104906

Quantity 5g

 View more versions of this product

Catalog No. AAH5104906

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 102308-32-7
    Molecular Formula C11H19NO4
    Formula Weight 229.27
    MDL Number MFCD00209557
    Beilstein 3591324
    Merck Index 14,4371
    Synonym (S)-(-)-3-tert-Butoxycarbonyl-2,2-dimethyloxazolidine-4-carboxaldehyde; Garner's Aldehyde
    Chemical Name or Material (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde

    Specifications

    Formula Weight 229.27
    CAS 102308-32-7
    Beilstein 3591324
    Merck Index 14,4371
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Optical Rotation −90° (c=1 in Chloroform)
    Recommended Storage 2° to 8°C
    Molecular Formula C11H19NO4
    MDL Number MFCD00209557
    Synonym (S)-(-)-3-tert-Butoxycarbonyl-2,2-dimethyloxazolidine-4-carboxaldehyde; Garner's Aldehyde
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 5g
    Chemical Name or Material (S)-(-)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
    Assay 95%

    Description

    Applications
    It is used to prepare γ-aminophosphonates (N-arylamide phosphonates as subtype-selective sphingosine 1-phosphate receptor agonists and antagonists. It is also used to synthesize N-sulfonylated propargylglycine derivatives as metalloproteinase inhibitors. Also widely utilized in the asymmetric synthesis of natural products.

    Solubility
    Slightly soluble in water.

    Notes
    Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit