Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid, 95%

Manufacturer:  Alfa Aesar™ H51987MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51987MD


  Check Availability
Add to cart

Specifications

Description

Applications
As lysine residue its 3-amino group is often used for carrying a tag moiety such as a fluorescent dye, a quencher or biotin.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
Description & Specifications

Chemical Identifiers

162558-25-0
426.47
7057792
(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid
C23H26N2O6
MFCD00235896
Nbeta-Boc-Nalpha-Fmoc-L-2,3-diaminopropionic acid; Fmoc-Dap(Boc)-OH

Specifications

162558-25-0
426.47
7057792
250mg
95%
C23H26N2O6
MFCD00235896
Nbeta-Boc-Nalpha-Fmoc-L-2,3-diaminopropionic acid; Fmoc-Dap(Boc)-OH
(S)-3-(Boc-amino)-2-(Fmoc-amino)propionic acid

Safety and Handling

SDS & Safety Documents

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit