Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid, 95%

Manufacturer:  Alfa Aesar™ H5219703

Quantity 1g

 View more versions of this product

Catalog No. AAH5219703


  Check Availability
Add to cart

Specifications

Description

Applications
(S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

219297-13-9
280.32
(S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid
C14H20N2O4
MFCD01076288

Specifications

219297-13-9
280.32
140° to 143°C
(S)-3-(Boc-amino)-4-(4-pyridyl)butyric acid
C14H20N2O4
MFCD01076288
1g
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit