Alfa Aesar™ (S)-3-(Boc-amino)adipic acid 6-benzyl ester, 95%

Manufacturer: Alfa Aesar™ H52187MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52187MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 218943-30-7
    Molecular Formula C18H25NO6
    Formula Weight 351.4
    MDL Number MFCD01862936
    Beilstein 8079168
    Synonym N-Boc-L-beta-homoglutamic acid 6-benzyl ester; Boc-beta-Homoglu(OBzl)-OH
    Chemical Name or Material (S)-3-(Boc-amino)adipic acid 6-benzyl ester

    Specifications

    Formula Weight 351.4
    Molecular Formula C18H25NO6
    MDL Number MFCD01862936
    CAS 218943-30-7
    Beilstein 8079168
    Synonym N-Boc-L-beta-homoglutamic acid 6-benzyl ester; Boc-beta-Homoglu(OBzl)-OH
    Quantity 250mg
    Chemical Name or Material (S)-3-(Boc-amino)adipic acid 6-benzyl ester
    Assay 95%

    Description

    Applications
    Starting materials for peptide synthesis, peptidomimetic and medicinal chemistry, protected amino acids, coupling reagents, linkers and resins, natural and unusual amino acids and building blocks used in peptide synthesis, peptidomimetic and medicinal chemistry. Reagents for PEGylation, Life science Research and biocatalysis.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit