Alfa Aesar™ (S)-3-(Boc-amino)butyric acid, 95%

Manufacturer: Alfa Aesar™ H52066MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52066MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 158851-30-0
    Molecular Formula C9H17NO4
    Formula Weight 203.24
    MDL Number MFCD00270345
    Beilstein 6800577
    Synonym N-Boc-L-beta-homoalanine; Boc-beta-Homoala-OH
    Chemical Name or Material (S)-3-(Boc-amino)butyric acid

    Specifications

    Formula Weight 203.24
    Molecular Formula C9H17NO4
    MDL Number MFCD00270345
    CAS 158851-30-0
    Beilstein 6800577
    Synonym N-Boc-L-beta-homoalanine; Boc-beta-Homoala-OH
    Quantity 250mg
    Chemical Name or Material (S)-3-(Boc-amino)butyric acid
    Assay 95%

    Description

    Applications
    (S)-3-(Boc-amino)butyric acid is used for the syntheses of aza-analogues of macrosphelides, synthesis of hybrid peptides with 12/10-helix, and in the synthesis of potentially pharmaceutically active products for the treatment of auto-immune and inflammatory diseases.

    Solubility
    Slightly soluble in water.

    Notes
    Stable under recommended storage conditions. Store away from oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit