Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Carboxy-4-hydroxyphenylglycine, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

55136-48-6
C9H9NO5
211.173
CHZBCZTXSTWCIG-SSDOTTSWSA-N
6604713

Group I antagonist/group II agonist

Mixed group I metabotropic glutamate receptor antagonist and group II mGlu agonist. (R)-3-Carboxy-4-hydroxyphenylglycine (Cat. No. 0328) also available.
CAS 55136-48-6
Molecular Formula C9H9NO5
InChI Key CHZBCZTXSTWCIG-SSDOTTSWSA-N
SMILES C1=CC(=C(C=C1C(C(=O)O)N)C(=O)O)O
IUPAC Name 5-[(R)-amino(carboxy)methyl]-2-hydroxybenzoic acid
Molecular Weight (g/mol) 211.173
PubChem CID 6604713
Formula Weight 211.17
Quantity 10mg
Chemical Name or Material (S)-3-Carboxy-4-hydroxyphenylglycine
Grade >98%
Show More Show Less

Recommended Storage : Store at Room Temperature