Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane, 98%, Acros Organics

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
60456-22-6
C6H11ClO2
150.61
MFCD00273365
BNPOTXLWPZOESZ-RXMQYKEDSA-N
s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane,4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane,s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane,s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane,1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s,s-3-chloro-1,2-propanediol acetonide,pubchem14076,bnpotxlwpzoesz-rxmqykedsa
6930483
(4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
CC1(OCC(O1)CCl)C
60456-22-6
C6H11ClO2
150.61
MFCD00273365
BNPOTXLWPZOESZ-RXMQYKEDSA-N
s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane,4s-4-chloromethyl-2,2-dimethyl-1,3-dioxolane,s---4-chloromethyl-2,2-dimethyl-1,3-dioxolane,s-+-4-chloromethyl-2,2-dimethyl-1,3-dioxolane,1,3-dioxolane, 4-chloromethyl-2,2-dimethyl-, s,s-3-chloro-1,2-propanediol acetonide,pubchem14076,bnpotxlwpzoesz-rxmqykedsa
6930483
(4S)-4-(chloromethyl)-2,2-dimethyl-1,3-dioxolane
CC1(OCC(O1)CCl)C
Specific Rotation -42
Specific Gravity 1.095
Refractive Index 1.4330 to 1.4350
Packaging Glass Bottle
Quantity 1g
Flash Point 50°C
Density 1.0950g/mL
Boiling Point 63.0°C (37.0 mmHg)
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: soluble
Formula Weight 150.61
Percent Purity 98%
Chemical Name or Material (S)-(-)-4-(Chloromethyl)-2,2-dimethyl-1,3-dioxolane
Show More Show Less

GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

TSCA : TSCA