Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid, 95%

Manufacturer:  Alfa Aesar™ H52101MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH52101MD


  Check Availability
Add to cart

Specifications

Description

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Description & Specifications

Chemical Identifiers

507472-25-5
432.43
N-Fmoc-2-nitro-D-beta-phenylalanine; Fmoc-D-beta-Phe(2-NO2)-OH
C24H20N2O6
MFCD03428007
(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid

Specifications

507472-25-5
432.43
N-Fmoc-2-nitro-D-beta-phenylalanine; Fmoc-D-beta-Phe(2-NO2)-OH
250mg
95%
C24H20N2O6
MFCD03428007
154° to 156°C
(S)-3-(Fmoc-amino)-3-(2-nitrophenyl)propionic acid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit