Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Hydroxyphenylglycine, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
71301-82-1
C8H9NO3
167.164
DQLYTFPAEVJTFM-ZETCQYMHSA-N
s-3-hydroxyphenylglycine,2s-2-amino-2-3-hydroxyphenyl acetic acid,s-2-3-hydroxyphenyl glycine,s-3hpg,s-3h-pg,s-amino-3-hydroxy-phenyl-acetic acid,s-2-amino-2-3-hydroxyphenyl acetic acid,3hpg,tocris-0324,tocris-0326
6604712
(2S)-2-amino-2-(3-hydroxyphenyl)acetic acid
C1=CC(=CC(=C1)O)C(C(=O)O)N
71301-82-1
C8H9NO3
167.164
DQLYTFPAEVJTFM-ZETCQYMHSA-N
s-3-hydroxyphenylglycine,2s-2-amino-2-3-hydroxyphenyl acetic acid,s-2-3-hydroxyphenyl glycine,s-3hpg,s-3h-pg,s-amino-3-hydroxy-phenyl-acetic acid,s-2-amino-2-3-hydroxyphenyl acetic acid,3hpg,tocris-0324,tocris-0326
6604712
(2S)-2-amino-2-(3-hydroxyphenyl)acetic acid
C1=CC(=CC(=C1)O)C(C(=O)O)N

Group I mGlu agonist, active isomer

Agonist at group I metabotropic glutamate receptors (mGlu1) having no effect at mGlu2 or mGlu4. (RS)-3-Hydroxyphenylglycine (Cat. No. 0324) also available.
Formula Weight 167.16
Percent Purity >99%
Quantity 50mg
Chemical Name or Material (S)-3-Hydroxyphenylglycine

Recommended Storage : Desiccate at Room Temperature