Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Methylmorpholine hydrochloride, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
1022094-03-6
C5H11NO·ClH
137.61
MSOCQCWIEBVSLF-JEDNCBNOSA-N
s-3-methylmorpholine hydrochloride,s-3-methylmorpholine hcl,3s-3-methylmorpholine hydrochloride,s-3-methyl-morpholine hydrochloride,s-3-methyl morpholine hydrochloride,c5h11no.clh,s-3-methylmorpholinehydrochloride,s-3-methylmorpholin hydrochloride,3s-3-methylmorpholine-hydrogen chloride 1/1
53486469
(3S)-3-methylmorpholine;hydrochloride
CC1COCCN1.Cl
1022094-03-6
C5H11NO·ClH
137.61
MSOCQCWIEBVSLF-JEDNCBNOSA-N
s-3-methylmorpholine hydrochloride,s-3-methylmorpholine hcl,3s-3-methylmorpholine hydrochloride,s-3-methyl-morpholine hydrochloride,s-3-methyl morpholine hydrochloride,c5h11no.clh,s-3-methylmorpholinehydrochloride,s-3-methylmorpholin hydrochloride,3s-3-methylmorpholine-hydrogen chloride 1/1
53486469
(3S)-3-methylmorpholine;hydrochloride
CC1COCCN1.Cl
Packaging Glass Bottle
Quantity 1g
Infrared Spectrum Authentic
Melting Point 121.0°C to 124.0°C
Assay Percent Range 96.0 to max. 104% min. (Argentometry)
Formula Weight 137.61
Percent Purity 97%
Chemical Name or Material (S)-3-Methylmorpholine hydrochloride

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning