Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid, 95%

Click to view available options
Quantity:
250mg
1g
339531-50-9
C27H32N2O6
480.561
MFCD03490501
BUYLKVIICSUHDX-GOTSBHOMSA-N
fmoc-leu-ser psi me,me pro-oh,fmoc-leu-ser psi me,me pro-oh,fmoc-l-leu-ser psime,me.pro-oh,fmoc-leu-ser psi me,me pro-oh,ambotzfaa5001,fmoc-l-leu-n,o-isopropylidene-l-ser-oh,4s-3-fmoc-leu-2,2-dimethyl-oxazolidine-4-carboxylic acid,4s-3-2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylpentanoyl-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid,4s-3-2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoyl-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
51340577
(4S)-3-[(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
CC(C)CC(C(=O)N1C(COC1(C)C)C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
339531-50-9
C27H32N2O6
480.561
MFCD03490501
BUYLKVIICSUHDX-GOTSBHOMSA-N
fmoc-leu-ser psi me,me pro-oh,fmoc-leu-ser psi me,me pro-oh,fmoc-l-leu-ser psime,me.pro-oh,fmoc-leu-ser psi me,me pro-oh,ambotzfaa5001,fmoc-l-leu-n,o-isopropylidene-l-ser-oh,4s-3-fmoc-leu-2,2-dimethyl-oxazolidine-4-carboxylic acid,4s-3-2s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-methylpentanoyl-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid,4s-3-2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoyl-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
51340577
(4S)-3-[(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoyl]-2,2-dimethyl-1,3-oxazolidine-4-carboxylic acid
CC(C)CC(C(=O)N1C(COC1(C)C)C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Quantity 1g
Formula Weight 480.56
Physical Form Solid
Percent Purity 95%
Chemical Name or Material (S)-3-(N-Fmoc-L-leucinyl)-2,2-dimethyloxazolidine-4-carboxylic acid