Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-3-Phenylcyclopentanone 98.0+%, TCI America™

Click to view available options
Quantity:
100mg
1g
86505-50-2
C11H12O
160.216
MFBXYJLOYZMFIN-JTQLQIEISA-N
10931730
(3S)-3-phenylcyclopentan-1-one
C1CC(=O)CC1C2=CC=CC=C2
86505-50-2
C11H12O
160.216
MFBXYJLOYZMFIN-JTQLQIEISA-N
10931730
(3S)-3-phenylcyclopentan-1-one
C1CC(=O)CC1C2=CC=CC=C2
Color Yellow
Boiling Point 155°C
Quantity 1g
Formula Weight 160.22
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material (S)-3-Phenylcyclopentanone