Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-3-Piperidinamine dihydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
334618-07-4
C5H12N2·2ClH
173.09
GGPNYXIOFZLNKW-XRIGFGBMSA-N
s-3-aminopiperidine dihydrochloride,s-piperidin-3-amine dihydrochloride,s-+-3-aminopiperidine dihydrochloride,s-3-aminopiperidine 2hcl,3s-piperidin-3-amine dihydrochloride,s-+-3-aminopiperidine 2hcl,3-piperidinamine, dihydrochloride, 3s,3s-3-piperidinamine dihydrochloride,3s-3-aminopiperidine dihydrochloride,s-piperidin-3-ylamine dihydrochloride
16218278
(3S)-piperidin-3-amine;dihydrochloride
C1CC(CNC1)N.Cl.Cl
334618-07-4
C5H12N2·2ClH
173.09
GGPNYXIOFZLNKW-XRIGFGBMSA-N
s-3-aminopiperidine dihydrochloride,s-piperidin-3-amine dihydrochloride,s-+-3-aminopiperidine dihydrochloride,s-3-aminopiperidine 2hcl,3s-piperidin-3-amine dihydrochloride,s-+-3-aminopiperidine 2hcl,3-piperidinamine, dihydrochloride, 3s,3s-3-piperidinamine dihydrochloride,3s-3-aminopiperidine dihydrochloride,s-piperidin-3-ylamine dihydrochloride
16218278
(3S)-piperidin-3-amine;dihydrochloride
C1CC(CNC1)N.Cl.Cl
Melting Point 217.2°C to 217.9°C
Color White
Infrared Spectrum Authentic
Quantity 5g
Specific Rotation +15° to +25° (20°C, 589nm) (c=1, water)
Elemental Analysis 33.31 to 36.09% C, 15.53 to 16.83% N
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 173.09
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (S)-(+)-3-Piperidinamine dihydrochloride
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a P

GHS Signal Word: Warning