Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-3-Pyrrolidinol, 98+%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
100243-39-8
C4H9NO
87.12
MFCD00192426
JHHZLHWJQPUNKB-BYPYZUCNSA-N
s-pyrrolidin-3-ol,s-3-hydroxypyrrolidine,3s-pyrrolidin-3-ol,s-3-pyrrolidinol,s---3-pyrrolidinol,3-pyrrolidinol, 3s,3s-3-pyrrolidinol,3s-3-hydroxypyrrolidine,s-3-hydroxy-pyrrolidine,s---3-hydroxypyrrolidine
2733874
(3S)-pyrrolidin-3-ol
C1CNCC1O
100243-39-8
C4H9NO
87.12
MFCD00192426
JHHZLHWJQPUNKB-BYPYZUCNSA-N
s-pyrrolidin-3-ol,s-3-hydroxypyrrolidine,3s-pyrrolidin-3-ol,s-3-pyrrolidinol,s---3-pyrrolidinol,3-pyrrolidinol, 3s,3s-3-pyrrolidinol,3s-3-hydroxypyrrolidine,s-3-hydroxy-pyrrolidine,s---3-hydroxypyrrolidine
2733874
(3S)-pyrrolidin-3-ol
C1CNCC1O
Boiling Point 108°C to 110°C (8.0mmHg)
Flash Point >110°C
Specific Gravity 1.048
Density 1.0480g/mL
Refractive Index 1.4870 to 1.491
Quantity 1g
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98% min. (GC)
Formula Weight 87.12
Physical Form Liquid After Melting
Percent Purity 98+%
Chemical Name or Material (S)-(-)-3-Pyrrolidinol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Recommended Storage : Product darkens on storage