Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S 32212 hydrochloride, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
847871-78-7
C25H29ClN4O2
452.983
RAKGNVNONBJBSW-UHFFFAOYSA-N
s 32212 hydrochloride,1,2-dihydro-n-4-methoxy-3-4-methyl-1-piperazinyl phenyl-3h-benz e indole-3-carboxamide hydrochloride
11487816
N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]-1,2-dihydrobenzo[e]indole-3-carboxamide;hydrochloride
CN1CCN(CC1)C2=C(C=CC(=C2)NC(=O)N3CCC4=C3C=CC5=CC=CC=C45)OC.Cl
This item is not returnable. View return policy
847871-78-7
C25H29ClN4O2
452.983
RAKGNVNONBJBSW-UHFFFAOYSA-N
s 32212 hydrochloride,1,2-dihydro-n-4-methoxy-3-4-methyl-1-piperazinyl phenyl-3h-benz e indole-3-carboxamide hydrochloride
11487816
N-[4-methoxy-3-(4-methylpiperazin-1-yl)phenyl]-1,2-dihydrobenzo[e]indole-3-carboxamide;hydrochloride
CN1CCN(CC1)C2=C(C=CC(=C2)NC(=O)N3CCC4=C3C=CC5=CC=CC=C45)OC.Cl

5-HT2C inverse agonist; α2 antagonist

Inverse agonist of 5-HT2C receptors (pKi = 8.18 at human 5-HT2C INI receptors). Also an antagonist of human α2-adrenoceptors. Displays negligible affinity for α1A-adrenoceptors, histamine H1 receptors and muscarinic M1 receptors.
Quantity 10mg
Formula Weight 452.98
Percent Purity >99%
Chemical Name or Material S 32212 hydrochloride

Recommended Storage : Desiccate at Room Temperature