Alfa Aesar™ (S)-(+)-4,12-Bis(di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS

Asymmetric hydrogenation

Manufacturer: Alfa Aesar™ 44616-77

Quantity 0.1g

 View more versions of this product

Catalog No. AA4461677

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 325168-88-5
    Molecular Formula C48H50P2
    Formula Weight 688.87
    MDL Number MFCD03840578
    Chemical Name or Material (S)- (+)-4,12-Bis (di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS

    Specifications

    Formula Weight 688.87
    CAS 325168-88-5
    Storage Note 1
    • Packaged under Argon
    Molecular Formula C48H50P2
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD03840578
    Color White
    Quantity 0.1g
    Chemical Name or Material (S)- (+)-4,12-Bis (di-3,5-xylylphosphino)[2.2]paracyclophane, CTH-(S)-3,5-xylyl-PHANEPHOS

    • Packaged under Argon

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit