Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid, 95%

Click to view available options
Quantity:
1g
62234-40-6
C12H16N2O4
252.27
MFCD00077904
KXMSCBRTBLPGIN-JTQLQIEISA-N
z-dab-oh,cbz-l-2,4-diaminobutyric acid,n-alpha-cbz-l-2,4-diamiobutyric acid,z-dbu-oh,s-4-amino-2-benzyloxy carbonyl amino butanoic acid,2s-4-amino-2-benzyloxy carbonyl amino butanoic acid,butanoic acid, 4-amino-2-phenylmethoxy carbonyl amino-, 2s,s-4-amino-2-benzyloxycarbonylamino butanoic acid,n-alpha-benzyloxycarbonyl-l-2,4-diaminobutyric acid,2s-4-amino-2-benzyloxy carbonyl amino butanoic acid
7018863
(2S)-4-amino-2-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCN)C(=O)O
62234-40-6
C12H16N2O4
252.27
MFCD00077904
KXMSCBRTBLPGIN-JTQLQIEISA-N
z-dab-oh,cbz-l-2,4-diaminobutyric acid,n-alpha-cbz-l-2,4-diamiobutyric acid,z-dbu-oh,s-4-amino-2-benzyloxy carbonyl amino butanoic acid,2s-4-amino-2-benzyloxy carbonyl amino butanoic acid,butanoic acid, 4-amino-2-phenylmethoxy carbonyl amino-, 2s,s-4-amino-2-benzyloxycarbonylamino butanoic acid,n-alpha-benzyloxycarbonyl-l-2,4-diaminobutyric acid,2s-4-amino-2-benzyloxy carbonyl amino butanoic acid
7018863
(2S)-4-amino-2-(phenylmethoxycarbonylamino)butanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCN)C(=O)O
Melting Point 194°C to 196°C
Quantity 1g
Color Brown
Beilstein 5960816
Formula Weight 252.27
Physical Form Powder
Percent Purity 95%
Chemical Name or Material (S)-4-Amino-2-(benzyloxycarbonylamino)butyric acid

storageNote1 : 2° to 8°C