Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(-)-4-Benzyl-2-oxazolidinone, 99%

Click to view available options
Quantity:
1g
5g
25g
90719-32-7
C10H11NO2
177.203
MFCD00064496
OJOFMLDBXPDXLQ-VIFPVBQESA-N
s-4-benzyl-2-oxazolidinone,s-4-benzyloxazolidin-2-one,4s-4-benzyl-1,3-oxazolidin-2-one,s---4-benzyl-2-oxazolidinone,s-4-benzyl-oxazolidin-2-one,2-oxazolidinone, 4-phenylmethyl-, 4s,s-+-4-benzyl-2-oxazolidinone,s-4-phenylmethyl-2-oxazolidinone,4s-4-phenylmethyl-1,3-oxazolidin-2-one
736225
(4S)-4-benzyl-1,3-oxazolidin-2-one
C1C(NC(=O)O1)CC2=CC=CC=C2
90719-32-7
C10H11NO2
177.203
MFCD00064496
OJOFMLDBXPDXLQ-VIFPVBQESA-N
s-4-benzyl-2-oxazolidinone,s-4-benzyloxazolidin-2-one,4s-4-benzyl-1,3-oxazolidin-2-one,s---4-benzyl-2-oxazolidinone,s-4-benzyl-oxazolidin-2-one,2-oxazolidinone, 4-phenylmethyl-, 4s,s-+-4-benzyl-2-oxazolidinone,s-4-phenylmethyl-2-oxazolidinone,4s-4-phenylmethyl-1,3-oxazolidin-2-one
736225
(4S)-4-benzyl-1,3-oxazolidin-2-one
C1C(NC(=O)O1)CC2=CC=CC=C2
Applications
(S)-(-)-4-Benzyl-2-oxazolidinone is used in chiral auxiliary asymmetric alkylation. It has been used in the synthesis of (S)-equol. Preparation and functionalization of chiral, stabilized ylides. It was used for asymmetric synthesis of (3R)- and (3S)-piperazic acid.

Solubility
Insoluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
Sensitivity Hygroscopic
Quantity 5g
Optical Rotation −64° (c=1 in Chloroform)
Melting Point 87°C to 89°C
Beilstein 3649667
Solubility Information Insoluble in water.
Formula Weight 177.2
Percent Purity 99%
Chemical Name or Material (S)-(-)-4-Benzyl-2-oxazolidinone

P261-P262-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P314-P330-P332+P313-P362-P501c

H302-H315-H320-H335-H373

TSCA : No

Recommended Storage : Ambient temperatures