Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-4-Boc-(3-hydroxymethyl)morpholine, 97%

Click to view available options
Quantity:
250mg
1g
5g
714971-28-5
C10H19NO4
217.265
MFCD06799478
AIQSXVGBMCJQAG-QMMMGPOBSA-N
s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate,s-4-boc-3-hydroxymethyl morpholine,s-n-boc-3-hydroxymethyl morpholine,tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate,s-n-boc-3-hydroxymethylmorpholine,3 s-hydroxymethyl-4-bocmorpholine,s-4-boc-3-hydroxymethylmorpholine,s-4-boc-3-hydroxymethyl-morpholine,s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester,pubchem5634
1514266
tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
CC(C)(C)OC(=O)N1CCOCC1CO
714971-28-5
C10H19NO4
217.265
MFCD06799478
AIQSXVGBMCJQAG-QMMMGPOBSA-N
s-tert-butyl 3-hydroxymethyl morpholine-4-carboxylate,s-4-boc-3-hydroxymethyl morpholine,s-n-boc-3-hydroxymethyl morpholine,tert-butyl 3s-3-hydroxymethyl morpholine-4-carboxylate,s-n-boc-3-hydroxymethylmorpholine,3 s-hydroxymethyl-4-bocmorpholine,s-4-boc-3-hydroxymethylmorpholine,s-4-boc-3-hydroxymethyl-morpholine,s-3-hydroxymethyl-morpholine-4-carboxylic acid tert-butyl ester,pubchem5634
1514266
tert-butyl (3S)-3-(hydroxymethyl)morpholine-4-carboxylate
CC(C)(C)OC(=O)N1CCOCC1CO
Applications
An intermediate of BMS-599626, a selective and orally efficacious inhibitor of human epidermal growth factor receptor 1 and 2 kinases.

Solubility
Slightly soluble in water.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Melting Point 80°C (literature)
Quantity 250mg
Color White
Beilstein 10251196
Formula Weight 217.27
Physical Form Powder
Percent Purity 97%
Chemical Name or Material (S)-4-Boc-(3-hydroxymethyl)morpholine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C