Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-4-Carboxyphenylglycine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

134052-73-6
C9H9NO4
195.174
VTMJKPGFERYGJF-ZETCQYMHSA-N
5311459

Competitive group I mGlu antagonist/weak group II agonist

Competitive group I metabotropic glutamate receptor antagonist, with selectivity for mGlu1a/1a over mGlu5a/5b. (RS)-4-Carboxyphenylglycine (Cat. No. 0321) and (R)-4-Carboxyphenylglycine (Cat. No. 0322) also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

CAS 134052-73-6
Molecular Formula C9H9NO4
InChI Key VTMJKPGFERYGJF-ZETCQYMHSA-N
SMILES C1=CC(=CC=C1C(C(=O)O)N)C(=O)O
IUPAC Name 4-[(S)-amino(carboxy)methyl]benzoic acid
Molecular Weight (g/mol) 195.174
PubChem CID 5311459
Formula Weight 195.17
Quantity 10mg
Chemical Name or Material (S)-4-Carboxyphenylglycine
Grade >99%
Show More Show Less

Recommended Storage : Store at Room Temperature