Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ S-(4-Chlorobenzyl)isothiouronium chloride, 98%

Manufacturer:  Alfa Aesar™ B2536214

Quantity 25g

 View more versions of this product

Catalog No. AAB2536214


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

544-47-8
237.15
14,2128
S-(4-Chlorobenzyl)isothiouronium chloride
C8H10Cl2N2S
MFCD00012584
2-(4-Chlorobenzyl)-2-thiopseudourea hydrochloride

Specifications

544-47-8
237.15
14,2128
202° to 206°C
S-(4-Chlorobenzyl)isothiouronium chloride
C8H10Cl2N2S
MFCD00012584
2-(4-Chlorobenzyl)-2-thiopseudourea hydrochloride
25g
98%

Safety and Handling

SDS & Safety Documents

RTECSNumber : UM0950000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit