Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(s)-(-)-4-fluoro-α-methylbenzyl alcohol, Acros Organics

Click to view available options
Quantity:
1g
101219-73-2
C8H9FO
140.157
PSDSORRYQPTKSV-LURJTMIESA-N
s-1-4-fluorophenyl ethanol,1s-1-4-fluorophenyl ethanol,1s-1-4-fluorophenyl ethan-1-ol,s-1-4-fluorophenyl-ethanol,benzenemethanol,4-fluoro-a-methyl-, as,pubchem14247,s-1-4-ffluorophenyl ethanol,s-1-4-fluorophenyl ethan-1-ol,1s-1-4-fluorophenyl ethan-1-o,s---1-4-fluorophenyl ethanol
852997
(1S)-1-(4-fluorophenyl)ethanol
CC(C1=CC=C(C=C1)F)O
101219-73-2
C8H9FO
140.157
PSDSORRYQPTKSV-LURJTMIESA-N
s-1-4-fluorophenyl ethanol,1s-1-4-fluorophenyl ethanol,1s-1-4-fluorophenyl ethan-1-ol,s-1-4-fluorophenyl-ethanol,benzenemethanol,4-fluoro-a-methyl-, as,pubchem14247,s-1-4-ffluorophenyl ethanol,s-1-4-fluorophenyl ethan-1-ol,1s-1-4-fluorophenyl ethan-1-o,s---1-4-fluorophenyl ethanol
852997
(1S)-1-(4-fluorophenyl)ethanol
CC(C1=CC=C(C=C1)F)O
Color Colorless
Infrared Spectrum Authentic
Enantiomeric Excess 97.5% min. (GC)
Quantity 1g
Linear Formula CH3CH(OH)C6H4F
Formula Weight 140.16
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material (S)-(-)-4-Fluoro-α-methylbenzyl alcohol