Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (S)-(-)-4-Hydroxy-2-pyrrolidinone, ≥97%

Click to view available options
Quantity:
250mg
1g
68108-18-9
C4H7NO2
101.105
MFCD00273370
IOGISYQVOGVIEU-VKHMYHEASA-N
s-4-hydroxy-2-pyrrolidinone,s-4-hydroxypyrrolidin-2-one,s-4-hydroxy-2-pyrrolidone,s-4-hydroxypyrrolidine-2-one,4s-4-hydroxypyrrolidin-2-one,s---4-hydroxy-2-pyrrolidinone,2-pyrrolidinone, 4-hydroxy-, 4s,s---4-hydroxy-2-pyrrolidone,s-4-hydroxy-pyrrolidine-2-one,4s-4-hydroxy-2-pyrrolidinone
155084
(4S)-4-hydroxypyrrolidin-2-one
C1C(CNC1=O)O
68108-18-9
C4H7NO2
101.105
MFCD00273370
IOGISYQVOGVIEU-VKHMYHEASA-N
s-4-hydroxy-2-pyrrolidinone,s-4-hydroxypyrrolidin-2-one,s-4-hydroxy-2-pyrrolidone,s-4-hydroxypyrrolidine-2-one,4s-4-hydroxypyrrolidin-2-one,s---4-hydroxy-2-pyrrolidinone,2-pyrrolidinone, 4-hydroxy-, 4s,s---4-hydroxy-2-pyrrolidone,s-4-hydroxy-pyrrolidine-2-one,4s-4-hydroxy-2-pyrrolidinone
155084
(4S)-4-hydroxypyrrolidin-2-one
C1C(CNC1=O)O
Color White
Melting Point 156°C to 159°C
Quantity 1g
Beilstein 1524192
Formula Weight 101.1
Physical Form Solid
Percent Purity ≥97%
Chemical Name or Material (S)-(-)-4-Hydroxy-2-pyrrolidinone