Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-4-Methyl-2-pentanol, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
14898-80-7
C6H14O
102.177
MFCD03093078
WVYWICLMDOOCFB-LURJTMIESA-N
s-+-4-methyl-2-pentanol,2s-4-methylpentan-2-ol,s-+-4-methylpentan-2-ol,4-methyl s-2-pentanol,s-4-methyl-pentan-2-ol,2s-4-methyl-2-pentanol,s-1,3-dimethyl-1-butanol,2-pentanol,4-methyl-, 2s,2-pentanol, 4-methyl-, 2s,unii-8u34xjk0r0 component
6994471
(2S)-4-methylpentan-2-ol
CC(C)CC(C)O
14898-80-7
C6H14O
102.177
MFCD03093078
WVYWICLMDOOCFB-LURJTMIESA-N
s-+-4-methyl-2-pentanol,2s-4-methylpentan-2-ol,s-+-4-methylpentan-2-ol,4-methyl s-2-pentanol,s-4-methyl-pentan-2-ol,2s-4-methyl-2-pentanol,s-1,3-dimethyl-1-butanol,2-pentanol,4-methyl-, 2s,2-pentanol, 4-methyl-, 2s,unii-8u34xjk0r0 component
6994471
(2S)-4-methylpentan-2-ol
CC(C)CC(C)O
Packaging Glass Bottle
Specific Rotation Condition +20° (20°C c=1,CHCl3)
Specific Rotation + 20.00
Specific Gravity 0.802
Refractive Index 1.4100 to 1.4120
Color Colorless
Density 0.8020g/mL
Boiling Point 132.0°C
Infrared Spectrum Authentic
Quantity 1g
Flash Point 41°C
Enantiomeric Excess 99.0% min.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 102.18
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-(+)-4-Methyl-2-pentanol
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning