Alfa Aesar™ (S)-(+)-4-Phenyl-2-oxazolidinone, 98%

Manufacturer: Alfa Aesar™ H2730606

Quantity 5g

 View more versions of this product

Catalog No. AAH2730606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 99395-88-7
    Molecular Formula C9H9NO2
    Formula Weight 163.18
    MDL Number MFCD00043396
    Chemical Name or Material (S)-(+)-4-Phenyl-2-oxazolidinone

    Specifications

    Formula Weight 163.18
    Molecular Formula C9H9NO2
    Quantity 5g
    Optical Rotation +52° (c=2 in Chloroform)
    MDL Number MFCD00043396
    CAS 99395-88-7
    Melting Point 130° to 132°C
    Chemical Name or Material (S)-(+)-4-Phenyl-2-oxazolidinone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit