Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-[(6-Chloro-2-oxooxazolo[4,5-b]pyridin-3yl)methyl] O,O-dimethyl phosphorothioate, 95%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
5g
35575-96-3
C9H10ClN2O5PS
324.672
MFCD00867611
VNKBTWQZTQIWDV-UHFFFAOYSA-N
azamethiphos,snip,ciba-geigy 18809,azamethiphos ban,azamethiophos,oms no 1825,6-chloro-3-dimethoxyphosphinoylthiomethyl-1,3-oxazolo 4,5-b pyridin-2 3h-one,s-6-chloro-2-oxooxazolo 4,5-b pyridin-3 2h-yl methyl o,o-dimethylphosphorothioate,s-6-chloro-2,3-dihydro-2-oxo-1,3-oxazolo 4,5-b pyridin-3-ylmethyl o,o-dimethyl phosphorothioate,azamethiphos ban
71482
CHEBI:38578
6-chloro-3-(dimethoxyphosphorylsulfanylmethyl)-[1,3]oxazolo[4,5-b]pyridin-2-one
COP(=O)(OC)SCN1C2=NC=C(C=C2OC1=O)Cl
35575-96-3
C9H10ClN2O5PS
324.672
MFCD00867611
VNKBTWQZTQIWDV-UHFFFAOYSA-N
azamethiphos,snip,ciba-geigy 18809,azamethiphos ban,azamethiophos,oms no 1825,6-chloro-3-dimethoxyphosphinoylthiomethyl-1,3-oxazolo 4,5-b pyridin-2 3h-one,s-6-chloro-2-oxooxazolo 4,5-b pyridin-3 2h-yl methyl o,o-dimethylphosphorothioate,s-6-chloro-2,3-dihydro-2-oxo-1,3-oxazolo 4,5-b pyridin-3-ylmethyl o,o-dimethyl phosphorothioate,azamethiphos ban
71482
CHEBI:38578
6-chloro-3-(dimethoxyphosphorylsulfanylmethyl)-[1,3]oxazolo[4,5-b]pyridin-2-one
COP(=O)(OC)SCN1C2=NC=C(C=C2OC1=O)Cl
Quantity 5g
Beilstein 1086470
Formula Weight 324.67
Physical Form Solid
Percent Purity 95%
Chemical Name or Material S-[(6-Chloro-2-oxooxazolo[4,5-b]pyridin-3yl)methyl] O,O-dimethyl phosphorothioate

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : TE8070000

storageNote1 : 2° to 8°C