Learn More
Azzota Corp S-63845 DFS Item
Manufacturer:  Azzota Corp C1370
S-63845 is a potent and selective MCL1 inhibitor (Ki (MCL1, FP) < 1.2nM; Kd (MCL1, SPR) = 0.19nM; Ki (BCL2, FP) > 10.000 1.2nM; Ki (BCL-XL, FP) > 10.000 1.2nM). S63845 specifically binds with high affinity to the BH3-binding groove of MCL1
Catalog No. 50-161-3203