Learn More
S-Adenosylmethionine Mouse, Clone: 118-18, Abnova™
Mouse monoclonal antibody raised against S-Adenosylmethionine.
Manufacturer:  Abnova Corporation MAB12271
Catalog No. 89-934-899
Add to cart
Description
Description
Specifications
Specifications
S-Adenosylmethionine
118-18
Mouse monoclonal antibody raised against S-Adenosylmethionine.
In 20mM PBS, 150mM NaCl, pH 7.4 (50% glycerol, 10mg/mL BSA, 0.02% sodium azide)
Mouse
IgG2b
25 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed)
Unconjugated
Competitive ELISA (1:4000-1:15000) Flow Cytometry (1:400) Immunohistochemistry (1:400) The optimal working dilution should be determined by the end user.
Liquid
S-Adenosylmethionine analog conjugated with KLH.
Affinity chromatography