Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S-Adenosylmethionine Mouse, Clone: 118-6, Abnova™
Mouse monoclonal antibody raised against S-Adenosylmethionine.
Supplier: Abnova Corporation MAB12269
Specifications
S-Adenosylmethionine | |
Monoclonal | |
Unconjugated | |
Competitive ELISA (1:10000) Flow Cytometry (1:200-400) Immunohistochemistry (1:200-400) The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity chromatography | |
RUO | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed) | |
118-6 | |
Mouse monoclonal antibody raised against S-Adenosylmethionine. | |
In PBS, pH 7.4 (50% glycerol, 1% BSA, 0.02% sodium azide) | |
S-Adenosylmethionine analog Aza-SAM conjugated with KLH. | |
25 μL | |
Primary | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction