Learn More
Accela Chembio IncSupplier Diversity Partner (S)-(-)-alpha-Amino-gamma-butyrolactone Hydrobromide, 15295-77-9, MFCD00674493, 5 g  DFS Item
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY0275015G
MF: C4H8BrNO2, MW: 182.02, ≥95%, BP: 281.4 celsius/760mmHg.
Catalog No. 50-650-109