(S)-(-)-Atenolol, Tocris

β1-antagonist; Active isomer of atenolol

Manufacturer: R&D Systems 039350

Quantity 50mg

 View more versions of this product

Catalog No. 03-935-0

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Assay Percent Range >97%
    Molecular Formula C14H22N2O3
    Formula Weight 266.34
    Synonym (S)-(-)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
    Chemical Name or Material (S)-(-)-Atenolol

    Specifications

    Quantity 50mg
    Formula Weight 266.34
    Assay Percent Range >97%
    Molecular Formula C14H22N2O3
    Recommended Storage Store at Room Temperature
    CAS 93379-54-5
    Synonym (S)-(-)-4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]benzeneacetamide
    Chemical Name or Material (S)-(-)-Atenolol

    Store at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit