Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-(-)-Bay K 8644, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
L-type Ca2+-channel activator with positive inotropic, vasoconstrictive and behavioral effects in vivo.
Racemate and (R)-(+)-Enantiomer also available.
Specifications
Quantity | 10 mg |
Synonym | s---bay k 8644, bay-k-8644 s---, --s-bayk8644, s---bay k8644, bay-k-8644 s--, methyl 4s-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-1,4-dihydropyridine-3-carboxylate, s---1,4-dihydro-2,6-dimethyl-5-nitro-4-2-trifluoromethyl phenyl-3-pyridinecarboxylic acid methyl ester, tocris-1544, lopac-b-112, d02rih |
Formula Weight | 356.3 |
Percent Purity | >99% |
Chemical Name or Material | (S)-(-)-Bay K 8644 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction