Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ S-Benzhydryl-N-Boc-L-cysteine, 98%

Click to view available options
Quantity:
250mg
1g
5g
21947-97-7
C21H25NO4S
387.494
MFCD00151869
AMTHPOXIVDHMLZ-KRWDZBQOSA-N
boc-cys dpm-oh,boc-s-diphenylmethyl-l-cysteine,2r-2-tert-butoxycarbonyl amino-3-diphenylmethyl sulfanyl propanoic acid,l-cysteine,n-1,1-dimethylethoxy carbonyl-s-diphenylmethyl
7015794
(2R)-3-benzhydrylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O
21947-97-7
C21H25NO4S
387.494
MFCD00151869
AMTHPOXIVDHMLZ-KRWDZBQOSA-N
boc-cys dpm-oh,boc-s-diphenylmethyl-l-cysteine,2r-2-tert-butoxycarbonyl amino-3-diphenylmethyl sulfanyl propanoic acid,l-cysteine,n-1,1-dimethylethoxy carbonyl-s-diphenylmethyl
7015794
(2R)-3-benzhydrylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O
Formula Weight 387.49
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material S-Benzhydryl-N-Boc-L-cysteine