Alfa Aesar™ (S)-Benzyl 1-[(S)-2-benzyloxycarbonylamino-6-(2,2,2-trifluoroacetamido)hexanoyl]pyrrolidine-2-carboxylate, 95%

Manufacturer: Alfa Aesar™ H3416003

Quantity 1g

 View more versions of this product

Catalog No. AAH3416003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114753-64-9
    Molecular Formula C28H32F3N3O6
    Formula Weight 563.57
    MDL Number MFCD22380709
    Synonym N2-Phenylmethoxycarbonyl-N6-(2,2,2-trifluoroacetyl)-L-lysyl-L-proline phenylmethyl ester
    Chemical Name or Material (S)-Benzyl 1-[(S)-2-benzyloxycarbonylamino-6-(2,2,2-trifluoroacetamido)hexanoyl]pyrrolidine-2-carboxylate

    Specifications

    Formula Weight 563.57
    Molecular Formula C28H32F3N3O6
    MDL Number MFCD22380709
    CAS 114753-64-9
    Synonym N2-Phenylmethoxycarbonyl-N6-(2,2,2-trifluoroacetyl)-L-lysyl-L-proline phenylmethyl ester
    Quantity 1g
    Chemical Name or Material (S)-Benzyl 1-[(S)-2-benzyloxycarbonylamino-6-(2,2,2-trifluoroacetamido)hexanoyl]pyrrolidine-2-carboxylate
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit