Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-beta-Methylphenethylamine 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
17596-79-1
C9H13N
135.21
MFCD00216741
AXORVIZLPOGIRG-UHFFFAOYNA-N
(S)-(-)-2-Phenyl-1-propylamine
1547950
2-phenylpropan-1-amine
CC(CN)C1=CC=CC=C1
17596-79-1
C9H13N
135.21
MFCD00216741
AXORVIZLPOGIRG-UHFFFAOYNA-N
(S)-(-)-2-Phenyl-1-propylamine
1547950
2-phenylpropan-1-amine
CC(CN)C1=CC=CC=C1
Color Yellow
Quantity 5g
UN Number 2735
Formula Weight 135.21
Physical Form Clear Liquid at 20°C
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material (S)-(-)-beta-Methylphenethylamine