Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-BOC-2,4-Dichlorophenylalanine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
114873-04-0
C14H17Cl2NO4
334.193
MFCD01862942
KSDWBXFRQWMWHO-NSHDSACASA-N
boc-l-2,4-dichlorophenylalanine,s-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid,s-boc-2,4-dichlorophenylalanine,boc-d-phe 2,4-cl2-oh,boc-2,4-dichloro-l-phenylalanin,2s-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid,l-phenylalanine, 2,4-dichloro-n-1,1-dimethylethoxy carbonyl,d-boc-2,4-dichlorophenylalanine,boc-phe 2,4-cl2-oh,l-boc-2,4-dichlorophenylalanine
2734683
(2S)-3-(2,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=C(C=C(C=C1)Cl)Cl)C(=O)O
114873-04-0
C14H17Cl2NO4
334.193
MFCD01862942
KSDWBXFRQWMWHO-NSHDSACASA-N
boc-l-2,4-dichlorophenylalanine,s-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid,s-boc-2,4-dichlorophenylalanine,boc-d-phe 2,4-cl2-oh,boc-2,4-dichloro-l-phenylalanin,2s-2-tert-butoxycarbonyl amino-3-2,4-dichlorophenyl propanoic acid,l-phenylalanine, 2,4-dichloro-n-1,1-dimethylethoxy carbonyl,d-boc-2,4-dichlorophenylalanine,boc-phe 2,4-cl2-oh,l-boc-2,4-dichlorophenylalanine
2734683
(2S)-3-(2,4-dichlorophenyl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)NC(CC1=C(C=C(C=C1)Cl)Cl)C(=O)O
Specific Rotation -30° to -34° (20°C, 589nm) (C=1, Methanol)
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 334.2
Physical Form Powder
Percent Purity ≥98.5%
Chemical Name or Material (S)-BOC-2, 4-Dichlorophenylalanine
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.