Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Butyrylthiocholine iodide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1866-16-6
C9H20INOS
317.22
WEQAAFZDJROSBF-UHFFFAOYSA-M
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
74630
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
CCCC(=O)SCC[N+](C)(C)C.[I-]
1866-16-6
C9H20INOS
317.22
WEQAAFZDJROSBF-UHFFFAOYSA-M
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
74630
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
CCCC(=O)SCC[N+](C)(C)C.[I-]
Color White
Packaging Glass bottle
Quantity 25g
Melting Point 172.0°C to 176.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3CH2CH2COSCH2CH2N(CH3)3I
Beilstein 04,IV,1585
Solubility Information Solubility in water: freely soluble
Formula Weight 317.22
Physical Form Crystals or Crystalline Powder
Percent Purity 98%
Chemical Name or Material S-Butyrylthiocholine iodide
Show More Show Less

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 217-475-7