Alfa Aesar™ S-Carboxymethyl-L-cysteine, 97%

Manufacturer: Alfa Aesar™ B2348722

Quantity 100g

 View more versions of this product

Catalog No. AAB2348722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 638-23-3
    Molecular Formula C5H9NO4S
    Formula Weight 179.2
    MDL Number MFCD00002614
    Beilstein 1725012
    Merck Index 14,1802
    Synonym Carbocysteine; 3-Carboxymethylthio-L-alanine
    Chemical Name or Material S-Carboxymethyl-L-cysteine

    Specifications

    Formula Weight 179.2
    TSCA TSCA
    Molecular Formula C5H9NO4S
    MDL Number MFCD00002614
    CAS 638-23-3
    Beilstein 1725012
    Merck Index 14,1802
    Synonym Carbocysteine; 3-Carboxymethylthio-L-alanine
    Melting Point ≈200°C (dec.)
    Quantity 100g
    Optical Rotation −29° (c=10 in 10% NaHCO)
    Recommended Storage 2° to 8°C
    RTECS Number AY4342000
    Chemical Name or Material S-Carboxymethyl-L-cysteine
    Assay 97%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit